Product Details
- SNP ID
-
rs192352490
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35765176 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGGCCTTGGCCCCAGGGCACCC[A/G]GTAAGGGCTGAGAGGCAAAGACTGA
- Phenotype
-
MIM: 610695
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HSPB6
PubMed Links
Gene Details
- Gene
- HSPB6
- Gene Name
- heat shock protein family B (small) member 6
There are no transcripts associated with this gene.
- Gene
- PROSER3
- Gene Name
- proline and serine rich 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039887.2 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
NP_001034976.2 |
XM_011526528.1 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
XP_011524830.1 |
XM_011526529.2 |
842 |
Missense Mutation |
ACA,GCA |
T256A |
XP_011524831.1 |
XM_011526530.1 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
XP_011524832.1 |
XM_011526531.1 |
842 |
Missense Mutation |
ACA,GCA |
T246A |
XP_011524833.1 |
XM_011526532.1 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
XP_011524834.1 |
XM_011526533.2 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
XP_011524835.1 |
XM_011526534.1 |
842 |
Missense Mutation |
ACA,GCA |
T257A |
XP_011524836.1 |
XM_011526535.2 |
842 |
Missense Mutation |
ACA,GCA |
T157A |
XP_011524837.1 |
XM_011526536.2 |
842 |
Missense Mutation |
ACA,GCA |
T245A |
XP_011524838.1 |
View Full Product Details