Product Details

SNP ID
rs192398187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43446541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACAGTGAAGGAAAGGTCTTCGC[A/G]GGAGAGTCAAAGCAGCCTGAGGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM267 PubMed Links

Gene Details

Gene
TMEM267
Gene Name
transmembrane protein 267
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022483.4 1712 Missense Mutation CCG,CTG P110L NP_071928.2
XM_005248333.4 1712 Missense Mutation CCG,CTG P110L XP_005248390.1
XM_005248335.3 1712 Missense Mutation CCG,CTG P110L XP_005248392.1
XM_006714486.3 1712 Missense Mutation CCG,CTG P110L XP_006714549.1
XM_011514074.2 1712 Missense Mutation CCG,CTG P110L XP_011512376.1
XM_011514075.2 1712 Missense Mutation CCG,CTG P110L XP_011512377.1

View Full Product Details