Product Details

SNP ID
rs192475887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133187890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACAGGTCCCACCTCTCCAAGAA[C/T]GAAACAGTTCATGGGATAGGATCAG
Phenotype
MIM: 604753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF268 PubMed Links

Gene Details

Gene
ZNF268
Gene Name
zinc finger protein 268
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165881.2 382 Nonsense Mutation CGA,TGA R18* NP_001159353.1
NM_001165882.2 382 UTR 5 NP_001159354.1
NM_001165883.1 382 Nonsense Mutation CGA,TGA R18* NP_001159355.1
NM_001165884.2 382 Intron NP_001159356.2
NM_001165885.1 382 Nonsense Mutation CGA,TGA R18* NP_001159357.1
NM_001165886.1 382 Intron NP_001159358.1
NM_001165887.1 382 Intron NP_001159359.1
NM_003415.2 382 Nonsense Mutation CGA,TGA R18* NP_003406.1
NM_152943.2 382 Nonsense Mutation CGA,TGA R18* NP_694422.2

View Full Product Details