Product Details

SNP ID
rs193123513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29566651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGGAAAGTGCGCGCCACGCTG[C/T]TCCCGGAGGCGGGCAGGTCCCCGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRR15 PubMed Links

Gene Details

Gene
PRR15
Gene Name
proline rich 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175887.2 1837 Missense Mutation CTC,TTC L108F NP_787083.1
XM_005249653.3 1837 Missense Mutation CTC,TTC L108F XP_005249710.1
XM_011515198.1 1837 Missense Mutation CTC,TTC L108F XP_011513500.1
XM_011515199.1 1837 Missense Mutation CTC,TTC L108F XP_011513501.1
XM_017011842.1 1837 Missense Mutation CTC,TTC L108F XP_016867331.1

View Full Product Details