Product Details

SNP ID
rs193162306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29059070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGCAACACTCGGCCCTTCCAG[A/G]CTGCTGTCCCACTGTCCTGGGGTAG
Phenotype
MIM: 612062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf31 PubMed Links

Gene Details

Gene
C22orf31
Gene Name
chromosome 22 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015370.1 467 Missense Mutation GCC,GTC A182V NP_056185.1
XM_005261491.3 467 Missense Mutation GCC,GTC A78V XP_005261548.1
XM_011530096.2 467 Missense Mutation GCC,GTC A141V XP_011528398.1
XM_017028741.1 467 Missense Mutation GCC,GTC A141V XP_016884230.1
Gene
ZNRF3
Gene Name
zinc and ring finger 3
There are no transcripts associated with this gene.

View Full Product Details