Product Details

SNP ID
rs192870761
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75891166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGTCACCGAGTGGTCTGACGC[A/G]CGCACCTCCCAGTAGTGGTGCCCGG
Phenotype
MIM: 611844
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 1204 Silent Mutation CGC,CGT R367R NP_001243053.1
NM_173547.3 1204 Silent Mutation CGC,CGT R389R NP_775818.2
XM_006721760.3 1204 Silent Mutation CGC,CGT R402R XP_006721823.1
XM_006721761.3 1204 Silent Mutation CGC,CGT R380R XP_006721824.1
XM_011524499.2 1204 Intron XP_011522801.1
XM_011524500.2 1204 Intron XP_011522802.1
XM_011524501.2 1204 Intron XP_011522803.1
XM_011524502.2 1204 Intron XP_011522804.1
XM_011524503.2 1204 Intron XP_011522805.1
XM_017024343.1 1204 Intron XP_016879832.1
XM_017024344.1 1204 UTR 3 XP_016879833.1

View Full Product Details