Product Details
- SNP ID
-
rs192959499
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:15560865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGACCACCAACTGGCGTCTTTAT[A/T]TACATATTAGAGCGGGTCAGCCAAT
- Phenotype
-
MIM: 604300
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HACL1
PubMed Links
Gene Details
- Gene
- HACL1
- Gene Name
- 2-hydroxyacyl-CoA lyase 1
There are no transcripts associated with this gene.
View Full Product Details