Product Details

SNP ID
rs192944969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30970713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTCGGTGTTGCATCTTTTTTGT[C/T]GTTTATTTTTTTTTAAGCCAACGGA
Phenotype
MIM: 611358
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPRXP4 PubMed Links

Gene Details

Gene
DPRXP4
Gene Name
divergent-paired related homeobox pseudogene 4
There are no transcripts associated with this gene.

Gene
RNF135
Gene Name
ring finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184992.1 987 Intron NP_001171921.1
NM_032322.3 987 Intron NP_115698.3
NM_197939.1 987 Intron NP_922921.1
XM_005258043.4 987 UTR 5 XP_005258100.1
XM_017025223.1 987 Intron XP_016880712.1

View Full Product Details