Product Details

SNP ID
rs192985600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35287003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACTGCATCAGGAGGTCTGATTG[G/T]CCACGGCTTTGGTTTAGGGACAGCC
Phenotype
MIM: 602912
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF6 PubMed Links

Gene Details

Gene
EIF6
Gene Name
eukaryotic translation initiation factor 6
There are no transcripts associated with this gene.

Gene
FAM83C
Gene Name
family with sequence similarity 83 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178468.5 1894 Silent Mutation GGA,GGC G592G NP_848563.1
Gene
FAM83C-AS1
Gene Name
FAM83C antisense RNA 1
There are no transcripts associated with this gene.

Gene
MMP24-AS1
Gene Name
MMP24 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details