Product Details

SNP ID
rs192742504
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66948054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCCCCACCAGGGGAGAGCCTA[A/G]GGCTGTGAGTCTGTTGGAGTGTTGT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TCF24 PubMed Links

Gene Details

Gene
TCF24
Gene Name
transcription factor 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193502.1 1328 Silent Mutation CCC,CCT P167P NP_001180431.1
XM_017012940.1 1328 Silent Mutation CCC,CCT P167P XP_016868429.1

View Full Product Details