Product Details

SNP ID
rs192764066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108524321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCATGCAAGCACTTACGCTCC[A/G]TATGTCTGCGGCAAAAGCATGGGGC
Phenotype
MIM: 611684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FICD PubMed Links

Gene Details

Gene
FICD
Gene Name
FIC domain containing
There are no transcripts associated with this gene.

Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014706.3 2479 Silent Mutation TAC,TAT Y903Y NP_055521.1
XM_005269241.4 2479 Silent Mutation TAC,TAT Y921Y XP_005269298.1
XM_011539026.2 2479 Silent Mutation TAC,TAT Y615Y XP_011537328.1

View Full Product Details