Product Details

SNP ID
rs2864778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149982719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGATCACACAAGCCCGGAGACAG[T/C]GCAGCTTCTCCATTGACTCTGGTCG
Phenotype
MIM: 611365
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LOC646903 PubMed Links

Gene Details

Gene
LOC646903
Gene Name
uncharacterized LOC646903
There are no transcripts associated with this gene.

Gene
TMEM183B
Gene Name
transmembrane protein 183B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079809.1 Intron NP_001073277.1

View Full Product Details