Product Details

SNP ID
rs475320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60626806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATTAAAACCACAATGAAATA[C/T]TACAGCAAACTTGCCAGAATGGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00301 PubMed Links

Gene Details

Gene
LINC00301
Gene Name
long intergenic non-protein coding RNA 301
There are no transcripts associated with this gene.

View Full Product Details