Product Details
- SNP ID
-
rs471738
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
46
- Location
-
Chr.1:150304001 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAATGAGGGGTTCAATGTAATACT[G/A]TTTGATCAGTGCTATAAAAGAGACA
- Phenotype
-
MIM: 611984
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPS21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs471657] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPS21
- Gene Name
- mitochondrial ribosomal protein S21
View Full Product Details