Product Details

SNP ID
rs6701280
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:84506376 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGGAGGCTAGCGCGACCCGACT[C/G]TTAAGTTTTCTTTTCCTCTCCAGAG
Phenotype
MIM: 600874
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNG5 PubMed Links

Gene Details

Gene
GNG5
Gene Name
G protein subunit gamma 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005274.2 76 Intron NP_005265.1
Gene
RPF1
Gene Name
ribosome production factor 1 homolog
There are no transcripts associated with this gene.

Gene
SPATA1
Gene Name
spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310156.1 76 UTR 5 NP_001297085.1
XM_011542514.2 76 UTR 5 XP_011540816.1
XM_011542515.2 76 UTR 5 XP_011540817.1
XM_011542516.1 76 UTR 5 XP_011540818.1
XM_011542517.1 76 UTR 5 XP_011540819.1
XM_011542519.2 76 UTR 5 XP_011540821.1
XM_011542520.2 76 UTR 5 XP_011540822.1
XM_011542521.2 76 UTR 5 XP_011540823.1
XM_011542522.2 76 UTR 5 XP_011540824.1

View Full Product Details