Product Details

SNP ID
rs6705067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134933621 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAATGTCAAATATCATAAATTCA[A/G]TGTTTGGATATGCTTGCTTAGACTG
Phenotype
MIM: 603862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNT2 PubMed Links

Gene Details

Gene
CCNT2
Gene Name
cyclin T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001241.3 Intron NP_001232.1
NM_001320748.1 Intron NP_001307677.1
NM_001320749.1 Intron NP_001307678.1
NM_058241.2 Intron NP_490595.1
XM_017005227.1 Intron XP_016860716.1
XM_017005228.1 Intron XP_016860717.1
XM_017005229.1 Intron XP_016860718.1
XM_017005230.1 Intron XP_016860719.1
XM_017005231.1 Intron XP_016860720.1
XM_017005232.1 Intron XP_016860721.1
XM_017005233.1 Intron XP_016860722.1

View Full Product Details