Product Details

SNP ID
rs6755780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54466353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTTGTGTAATTAAGTATGTTTA[G/T]TCTTCGAGACCATCCTGGCTAACAC
Phenotype
MIM: 182790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPTBN1 PubMed Links

Gene Details

Gene
SPTBN1
Gene Name
spectrin beta, non-erythrocytic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003128.2 Intron NP_003119.2
NM_178313.2 Intron NP_842565.2
XM_005264517.2 Intron XP_005264574.1
XM_005264518.1 Intron XP_005264575.1
XM_006712087.2 Intron XP_006712150.1
XM_017004779.1 Intron XP_016860268.1
XM_017004780.1 Intron XP_016860269.1
XM_017004781.1 Intron XP_016860270.1

View Full Product Details