Product Details

SNP ID
rs6740465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218884873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGGAATATCTGGGTGCTGACCCC[C/T]TTTCTGACCCATGTTATCAATGGGT
Phenotype
MIM: 606268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT10A PubMed Links

Gene Details

Gene
WNT10A
Gene Name
Wnt family member 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025216.2 Intron NP_079492.2
XM_011511929.2 Intron XP_011510231.1
XM_011511930.1 Intron XP_011510232.1

View Full Product Details