Product Details
- SNP ID
-
rs7184856
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:86580611 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAATACACATGCAGCTATGTAA[C/G]CCACAACAGCAGACGTCCTATCCTT
- Phenotype
-
MIM: 603252
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXL1
PubMed Links
Gene Details
- Gene
- FOXL1
- Gene Name
- forkhead box L1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005250.2 |
2103 |
UTR 3 |
|
|
NP_005241.1 |
View Full Product Details