Product Details

SNP ID
rs7202498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57259073 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCAAGAGGCCACAGTTTCCAGGG[A/G]GTCAGCCCATGAGGCCCTGGTCCCC
Phenotype
MIM: 615407 MIM: 600340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL2BP PubMed Links

Gene Details

Gene
ARL2BP
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
There are no transcripts associated with this gene.

Gene
PLLP
Gene Name
plasmolipin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015993.2 Intron NP_057077.1

View Full Product Details