Product Details

SNP ID
rs7224127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30974590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGGGTCTATAATTCCTTCAGTG[A/G]TTTTTTTTTTTGTAGTTTTAAGTGT
Phenotype
MIM: 611358
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPRXP4 PubMed Links

Gene Details

Gene
DPRXP4
Gene Name
divergent-paired related homeobox pseudogene 4
There are no transcripts associated with this gene.

Gene
RNF135
Gene Name
ring finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184992.1 Intron NP_001171921.1
NM_032322.3 Intron NP_115698.3
NM_197939.1 Intron NP_922921.1
XM_005258043.4 Intron XP_005258100.1
XM_017025223.1 Intron XP_016880712.1

View Full Product Details