Product Details

SNP ID
rs7223247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1705735 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGTTTTTGTTTTGTTTTGTTTT[G/T]TTTTGAAGACAATCTTCCTGTCTCT
Phenotype
MIM: 612077
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR22 PubMed Links

Gene Details

Gene
MIR22
Gene Name
microRNA 22
There are no transcripts associated with this gene.

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
TLCD2
Gene Name
TLC domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164407.1 2956 UTR 3 NP_001157879.1

View Full Product Details