Product Details
- SNP ID
-
rs7238437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:3069519 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATTAAGAAATCTGTTTTTCTCCC[A/G]TCACTAATTAATATTTCTTACTGAA
- Phenotype
-
MIM: 603508
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYOM1
PubMed Links
Gene Details
- Gene
- MYOM1
- Gene Name
- myomesin 1
View Full Product Details