Product Details

SNP ID
rs748697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131103965 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTCCCCAGCTATTGAACAAGGA[G/A]GGCAAGCTGAACGCTGTGAGGAGTC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
AIF1L PubMed Links

Gene Details

Gene
AIF1L
Gene Name
allograft inflammatory factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185095.1 Intron NP_001172024.1
NM_001185096.1 Intron NP_001172025.1
NM_031426.3 Intron NP_113614.1
XM_017015177.1 Intron XP_016870666.1

View Full Product Details