Product Details

SNP ID
rs7592763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227166273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCATACATTAGGAATTCCCAG[A/C]CCAGGTATTTAAATGCTACTAGGCA
Phenotype
MIM: 120070 MIM: 120131
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COL4A3 PubMed Links

Gene Details

Gene
COL4A3
Gene Name
collagen type IV alpha 3 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000091.4 Intron NP_000082.2
XM_005246277.3 Intron XP_005246334.1
XM_005246280.3 Intron XP_005246337.1
XM_006712245.3 Intron XP_006712308.1
XM_011510555.1 Intron XP_011508857.1
XM_011510556.2 Intron XP_011508858.1
XM_017003295.1 Intron XP_016858784.1
Gene
COL4A4
Gene Name
collagen type IV alpha 4 chain
There are no transcripts associated with this gene.

View Full Product Details