Product Details

SNP ID
rs7723510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53884992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTAAAAAAAAAAAGCTTAAAG[A/C]AGTGTTTGATGTTTCAAAGGTAATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARL15 PubMed Links

Gene Details

Gene
ARL15
Gene Name
ADP ribosylation factor like GTPase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019087.2 2370 UTR 3 NP_061960.1
XM_011543498.2 2370 UTR 3 XP_011541800.1
XM_011543499.2 2370 UTR 3 XP_011541801.1
XM_011543500.2 2370 UTR 3 XP_011541802.1
XM_017009598.1 2370 UTR 3 XP_016865087.1

View Full Product Details