Product Details

SNP ID
rs7675050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112156483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTGCCCTCTCCACAGGTACAC[G/T]CTAAAACATAATCCCCATGAGGACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C4orf32 PubMed Links

Gene Details

Gene
C4orf32
Gene Name
chromosome 4 open reading frame 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152400.2 Intron NP_689613.1
XM_017007732.1 Intron XP_016863221.1

View Full Product Details