Product Details

SNP ID
rs7735019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:97162543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCACAAAACAGATTATTAATC[A/C]ATTAATTAAGCAAATTGTGCATCAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RIOK2 PubMed Links

Gene Details

Gene
RIOK2
Gene Name
RIO kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159749.1 2287 Intron NP_001153221.1
NM_018343.2 2287 UTR 3 NP_060813.2
XM_017009628.1 2287 UTR 3 XP_016865117.1

View Full Product Details