Product Details

SNP ID
rs7830133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116950310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTCACCTCTCAATAGGAGATC[A/C]GTTAATGCATACTGAAGGAAGGCTT
Phenotype
MIM: 611145
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AARD PubMed Links

Gene Details

Gene
AARD
Gene Name
alanine and arginine rich domain containing protein
There are no transcripts associated with this gene.

Gene
SLC30A8
Gene Name
solute carrier family 30 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172811.1 38 UTR 5 NP_001166282.1
NM_001172813.1 38 Intron NP_001166284.1
NM_001172814.1 38 Intron NP_001166285.1
NM_001172815.2 38 Intron NP_001166286.1
NM_173851.2 38 Intron NP_776250.2

View Full Product Details