Product Details

SNP ID
rs7827048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37798665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCAGTGGCAACCGTTTCTCCAC[C/T]GCCCCTTGCCCGCAGTCGGGGCTGG
Phenotype
MIM: 606823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRA2 PubMed Links

Gene Details

Gene
ADGRA2
Gene Name
adhesion G protein-coupled receptor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032777.9 Intron NP_116166.9
XM_005273471.3 Intron XP_005273528.1
XM_011544481.2 Intron XP_011542783.1
XM_011544482.2 Intron XP_011542784.1
XM_011544483.2 Intron XP_011542785.1

View Full Product Details