Product Details

SNP ID
rs7966050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122981330 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCACAGGCATCGGGGAGAGGAGG[C/T]GCAGTTACTACCCGGGGAAGTCGGG
Phenotype
MIM: 605453 MIM: 607668 MIM: 608920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB9 PubMed Links

Gene Details

Gene
ABCB9
Gene Name
ATP binding cassette subfamily B member 9
There are no transcripts associated with this gene.

Gene
ARL6IP4
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002251.2 Intron NP_001002251.2
NM_001002252.2 Intron NP_001002252.2
NM_001278378.1 Intron NP_001265307.1
NM_001278379.1 Intron NP_001265308.1
NM_001278380.1 Intron NP_001265309.1
NM_016638.3 Intron NP_057722.3
NM_018694.3 Intron NP_061164.3
Gene
OGFOD2
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
There are no transcripts associated with this gene.

Gene
PITPNM2
Gene Name
phosphatidylinositol transfer protein membrane associated 2
There are no transcripts associated with this gene.

View Full Product Details