Product Details

SNP ID
rs201349500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:130136499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGGCAGTTTGAGGAGCTGCTG[C/T]GCCTCAAAGCCAAGTAAGCGGGGCG
Phenotype
MIM: 612754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRX3 PubMed Links

Gene Details

Gene
GLRX3
Gene Name
glutaredoxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199868.1 91 Missense Mutation CGC,TGC R27C NP_001186797.1
NM_001321980.1 91 Intron NP_001308909.1
NM_006541.4 91 Intron NP_006532.2
XM_017015474.1 91 UTR 5 XP_016870963.1
Gene
LOC102724883
Gene Name
uncharacterized LOC102724883
There are no transcripts associated with this gene.

View Full Product Details