Product Details

SNP ID
rs200996195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116591780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCTTTCTCTGTAGGAAAAGAAG[A/T]TTGCTATGAGGACCTCGGGTGCTTT
Phenotype
MIM: 604422
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PNLIPRP1 PubMed Links

Gene Details

Gene
PNLIPRP1
Gene Name
pancreatic lipase related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303135.1 86 Missense Mutation GAT,GTT D20V NP_001290064.1
NM_006229.3 86 Missense Mutation GAT,GTT D20V NP_006220.1
XM_011539868.1 86 Missense Mutation GAT,GTT D20V XP_011538170.1
XM_011539869.1 86 UTR 5 XP_011538171.1

View Full Product Details