Product Details

SNP ID
rs201201168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62805226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGAACCTGAGCAAGCTGAAGA[A/G]CCTCCTGACCCAGCTCCGCGCCGAG
Phenotype
MIM: 611392 MIM: 129010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADO PubMed Links

Gene Details

Gene
ADO
Gene Name
2-aminoethanethiol dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032804.5 471 Missense Mutation AAC,AGC N56S NP_116193.2
Gene
EGR2
Gene Name
early growth response 2
There are no transcripts associated with this gene.

View Full Product Details