Product Details

SNP ID
rs200543036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122697913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATTTCGGTTTGTTAGGCCTGC[A/G]ATGGACCGTTCCGGTTTCTTTGCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 860 Silent Mutation ATC,ATT I276I NP_001010912.1

View Full Product Details