Product Details

SNP ID
rs200238542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:52316577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAAGAAGGTTCTGTTTGTCTC[C/T]GGTCATCAGACTGTGCCTCAGGATT
Phenotype
MIM: 605189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKK1 PubMed Links

Gene Details

Gene
DKK1
Gene Name
dickkopf WNT signaling pathway inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012242.2 725 Missense Mutation CGG,TGG R191W NP_036374.1
Gene
PRKG1-AS1
Gene Name
PRKG1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details