Product Details

SNP ID
rs200260532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21534788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTGCGACGGGCACGGCTGCAG[C/T]GTCGCGGTGCATCAAGGTAAACACC
Phenotype
MIM: 602409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MLLT10 PubMed Links

Gene Details

Gene
MLLT10
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195626.1 493 Silent Mutation AGC,AGT S48S NP_001182555.1
NM_001195627.1 493 Silent Mutation AGC,AGT S48S NP_001182556.1
NM_001195628.1 493 Silent Mutation AGC,AGT S48S NP_001182557.1
NM_001195630.1 493 Silent Mutation AGC,AGT S48S NP_001182559.1
NM_001324296.1 493 Silent Mutation AGC,AGT S48S NP_001311225.1
NM_001324297.1 493 UTR 5 NP_001311226.1
NM_004641.3 493 Silent Mutation AGC,AGT S48S NP_004632.1
Gene
SKIDA1
Gene Name
SKI/DACH domain containing 1
There are no transcripts associated with this gene.

View Full Product Details