Product Details

SNP ID
rs200476924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:61410495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACTGACTTCTCAACGAAGCAC[A/G]GACTGCCAATGCCAGCACCACCAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM26 PubMed Links

Gene Details

Gene
TMEM26
Gene Name
transmembrane protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178505.7 1303 Missense Mutation CGT,TGT R312C NP_848600.2
XM_011539450.2 1303 Intron XP_011537752.1
XM_011539451.1 1303 Intron XP_011537753.1
XM_017015829.1 1303 Intron XP_016871318.1

View Full Product Details