Product Details

SNP ID
rs200552223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110298656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTCTACACTTACTGTCCATC[C/T]TCACTCCAGACTGCCATACACTTGT
Phenotype
MIM: 603519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMNDC1 PubMed Links

Gene Details

Gene
SMNDC1
Gene Name
survival motor neuron domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005871.3 457 Silent Mutation GAA,GAG E85E NP_005862.1
XM_005269382.3 457 Silent Mutation GAA,GAG E46E XP_005269439.1

View Full Product Details