Product Details

SNP ID
rs200796165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72899022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGACCAAGCCCAGCGTCTGCTT[C/T]CACGTCTACTGTGGTCGTGAGTACC
Phenotype
MIM: 609330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OIT3 PubMed Links

Gene Details

Gene
OIT3
Gene Name
oncoprotein induced transcript 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152635.2 663 Silent Mutation TTC,TTT F140F NP_689848.1

View Full Product Details