Product Details

SNP ID
rs200786824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:28681293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTACTGTGATGCTGCCCACTGT[A/G]TAGCCACTGGTTATATGTGTAAATC
Phenotype
MIM: 604444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAMBI PubMed Links

Gene Details

Gene
BAMBI
Gene Name
BMP and activin membrane bound inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012342.2 515 Missense Mutation ATA,GTA I38V NP_036474.1

View Full Product Details