Product Details

SNP ID
rs200719626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97859530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTTTATCCAGAGAGACTACAGC[A/G]ATGGGACCATCTGTCAGTTCCAGAC
Phenotype
MIM: 606276 MIM: 614189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRTAC1 PubMed Links

Gene Details

Gene
CRTAC1
Gene Name
cartilage acidic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206528.2 151 Intron NP_001193457.1
NM_018058.6 151 Intron NP_060528.3
XM_005269938.4 151 Intron XP_005269995.1
XM_011539917.1 151 Intron XP_011538219.1
XM_017016366.1 151 Intron XP_016871855.1
XM_017016367.1 151 Intron XP_016871856.1
Gene
GOLGA7B
Gene Name
golgin A7 family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010917.2 151 Missense Mutation AAT,GAT N29D NP_001010917.1
Gene
LINC00866
Gene Name
long intergenic non-protein coding RNA 866
There are no transcripts associated with this gene.

View Full Product Details