Product Details

SNP ID
rs199922239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102403964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCATGGCCTTCAGCTTGGCCT[A/C]GTGGGTCCGCACCTGCTCCTCCTAG
Phenotype
MIM: 164012 MIM: 602327
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NFKB2 PubMed Links

Gene Details

Gene
NFKB2
Gene Name
nuclear factor kappa B subunit 2
There are no transcripts associated with this gene.

Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270965.1 2931 Nonsense Mutation GAG,TAG E908* NP_001257894.1
NM_001270966.1 2931 Nonsense Mutation GAG,TAG E529* NP_001257895.1
NM_002779.4 2931 Nonsense Mutation GAG,TAG E908* NP_002770.3
XM_011539968.1 2931 Nonsense Mutation GAG,TAG E908* XP_011538270.1
XM_011539969.2 2931 Nonsense Mutation GAG,TAG E908* XP_011538271.1
XM_017016433.1 2931 Nonsense Mutation GAG,TAG E908* XP_016871922.1
XM_017016434.1 2931 Nonsense Mutation GAG,TAG E908* XP_016871923.1

View Full Product Details