Product Details
- SNP ID
-
rs200144829
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:104132179 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGAATACTAATCAGAGCCTCATA[A/G]TTTGGTTCATTTAGGTACTTTGAGA
- Phenotype
-
MIM: 616527
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP43
PubMed Links
Gene Details
- Gene
- CFAP43
- Gene Name
- cilia and flagella associated protein 43
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025145.5 |
4350 |
Silent Mutation |
AAC,AAT |
N1538N |
NP_079421.5 |
XM_005270171.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1539N |
XP_005270228.1 |
XM_005270172.3 |
4350 |
Silent Mutation |
AAC,AAT |
N1510N |
XP_005270229.1 |
XM_011540196.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1577N |
XP_011538498.1 |
XM_011540197.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1548N |
XP_011538499.1 |
XM_011540198.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1538N |
XP_011538500.1 |
XM_011540199.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1538N |
XP_011538501.1 |
XM_011540200.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1411N |
XP_011538502.1 |
XM_011540201.2 |
4350 |
Intron |
|
|
XP_011538503.1 |
XM_011540202.2 |
4350 |
Silent Mutation |
AAC,AAT |
N1320N |
XP_011538504.1 |
XM_011540203.2 |
4350 |
Silent Mutation |
AAC,AAT |
N838N |
XP_011538505.1 |
XM_017016681.1 |
4350 |
Silent Mutation |
AAC,AAT |
N1576N |
XP_016872170.1 |
XM_017016682.1 |
4350 |
Silent Mutation |
AAC,AAT |
N1461N |
XP_016872171.1 |
XM_017016683.1 |
4350 |
Silent Mutation |
AAC,AAT |
N1039N |
XP_016872172.1 |
XM_017016684.1 |
4350 |
Intron |
|
|
XP_016872173.1 |
- Gene
- SFR1
- Gene Name
- SWI5 dependent homologous recombination repair protein 1
There are no transcripts associated with this gene.
View Full Product Details