Product Details
- SNP ID
-
rs200197026
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:45616499 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCCTCGTGCAAGTATCTCCTCCC[G/T]GCACTCTTGTAATCATAGGTACAGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZFAND4
PubMed Links
Gene Details
- Gene
- ZFAND4
- Gene Name
- zinc finger AN1-type containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128324.2 |
2823 |
Silent Mutation |
GCA,GCC |
A707A |
NP_001121796.1 |
NM_001282905.1 |
2823 |
Silent Mutation |
GCA,GCC |
A633A |
NP_001269834.1 |
NM_001282906.1 |
2823 |
Silent Mutation |
GCA,GCC |
A633A |
NP_001269835.1 |
NM_174890.3 |
2823 |
Silent Mutation |
GCA,GCC |
A707A |
NP_777550.2 |
XM_005271842.2 |
2823 |
Silent Mutation |
GCA,GCC |
A608A |
XP_005271899.1 |
XM_005271843.2 |
2823 |
Silent Mutation |
GCA,GCC |
A608A |
XP_005271900.1 |
XM_005271844.3 |
2823 |
Silent Mutation |
GCA,GCC |
A589A |
XP_005271901.1 |
XM_011540366.2 |
2823 |
Silent Mutation |
GCA,GCC |
A715A |
XP_011538668.1 |
XM_011540368.1 |
2823 |
Silent Mutation |
GCA,GCC |
A535A |
XP_011538670.1 |
XM_017016931.1 |
2823 |
Silent Mutation |
GCA,GCC |
A721A |
XP_016872420.1 |
XM_017016932.1 |
2823 |
Silent Mutation |
GCA,GCC |
A713A |
XP_016872421.1 |
XM_017016933.1 |
2823 |
Silent Mutation |
GCA,GCC |
A713A |
XP_016872422.1 |
XM_017016934.1 |
2823 |
Silent Mutation |
GCA,GCC |
A713A |
XP_016872423.1 |
XM_017016935.1 |
2823 |
Intron |
|
|
XP_016872424.1 |
XM_017016936.1 |
2823 |
Silent Mutation |
GCA,GCC |
A665A |
XP_016872425.1 |
XM_017016937.1 |
2823 |
Silent Mutation |
GCA,GCC |
A665A |
XP_016872426.1 |
XM_017016938.1 |
2823 |
Silent Mutation |
GCA,GCC |
A659A |
XP_016872427.1 |
XM_017016939.1 |
2823 |
Silent Mutation |
GCA,GCC |
A659A |
XP_016872428.1 |
XM_017016940.1 |
2823 |
Silent Mutation |
GCA,GCC |
A639A |
XP_016872429.1 |
XM_017016941.1 |
2823 |
Silent Mutation |
GCA,GCC |
A639A |
XP_016872430.1 |
XM_017016942.1 |
2823 |
Silent Mutation |
GCA,GCC |
A614A |
XP_016872431.1 |
XM_017016943.1 |
2823 |
Silent Mutation |
GCA,GCC |
A614A |
XP_016872432.1 |
XM_017016944.1 |
2823 |
Silent Mutation |
GCA,GCC |
A614A |
XP_016872433.1 |
XM_017016945.1 |
2823 |
Silent Mutation |
GCA,GCC |
A595A |
XP_016872434.1 |
XM_017016946.1 |
2823 |
Silent Mutation |
GCA,GCC |
A514A |
XP_016872435.1 |
View Full Product Details