Product Details

SNP ID
rs202111230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133309415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGACGTGCAGCGCCACCTCT[A/G]CCTCCAGGACGTGATCATGCAGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBGCP2 PubMed Links

Gene Details

Gene
TUBGCP2
Gene Name
tubulin gamma complex associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256617.1 234 Intron NP_001243546.1
NM_001256618.1 234 Intron NP_001243547.1
NM_006659.3 234 Intron NP_006650.1
XM_017015595.1 234 Intron XP_016871084.1
XM_017015596.1 234 Intron XP_016871085.1
Gene
ZNF511
Gene Name
zinc finger protein 511
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145806.3 234 Missense Mutation TAC,TGC Y60C NP_665805.2

View Full Product Details