Product Details

SNP ID
rs201881239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73911932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACTAGATACGAACAGGGTGAGG[C/T]GAGAGGGAGAGGGAAGAGTGGGTTT
Phenotype
MIM: 191840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf55 PubMed Links

Gene Details

Gene
C10orf55
Gene Name
chromosome 10 open reading frame 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001791.2 550 Missense Mutation NP_001001791.2
Gene
PLAU
Gene Name
plasminogen activator, urokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145031.2 550 Intron NP_001138503.1
NM_001319191.1 550 Intron NP_001306120.1
NM_002658.4 550 Intron NP_002649.1
XM_011539866.2 550 Intron XP_011538168.1

View Full Product Details