Product Details

SNP ID
rs201836632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102403968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCCTTCAGCTTGGCCTCGTG[G/T]GTCCGCACCTGCTCCTCCTAGCGGC
Phenotype
MIM: 164012 MIM: 602327
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NFKB2 PubMed Links

Gene Details

Gene
NFKB2
Gene Name
nuclear factor kappa B subunit 2
There are no transcripts associated with this gene.

Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270965.1 2927 Silent Mutation ACA,ACC T906T NP_001257894.1
NM_001270966.1 2927 Silent Mutation ACA,ACC T527T NP_001257895.1
NM_002779.4 2927 Silent Mutation ACA,ACC T906T NP_002770.3
XM_011539968.1 2927 Silent Mutation ACA,ACC T906T XP_011538270.1
XM_011539969.2 2927 Silent Mutation ACA,ACC T906T XP_011538271.1
XM_017016433.1 2927 Silent Mutation ACA,ACC T906T XP_016871922.1
XM_017016434.1 2927 Silent Mutation ACA,ACC T906T XP_016871923.1

View Full Product Details