Product Details
- SNP ID
-
rs201421429
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:37952135 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTTCCCAACTCATCTTACTTCT[C/G]AGCATTCCTCTTCTTTGTGTGTGTC
- Phenotype
-
MIM: 194528
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF25
PubMed Links
Gene Details
- Gene
- ZNF25
- Gene Name
- zinc finger protein 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_005252384.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q455E |
XP_005252441.1 |
XM_005252385.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q455E |
XP_005252442.1 |
XM_005252386.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q455E |
XP_005252443.1 |
XM_011519352.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q459E |
XP_011517654.1 |
XM_011519353.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q459E |
XP_011517655.1 |
XM_011519354.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q459E |
XP_011517656.1 |
XM_011519355.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q459E |
XP_011517657.1 |
XM_011519356.2 |
1706 |
Missense Mutation |
CAG,GAG |
Q419E |
XP_011517658.1 |
XM_017015837.1 |
1706 |
Missense Mutation |
CAG,GAG |
Q419E |
XP_016871326.1 |
XM_017015838.1 |
1706 |
Missense Mutation |
CAG,GAG |
Q419E |
XP_016871327.1 |
View Full Product Details