Product Details

SNP ID
rs201588650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96368094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCAAGAACATCAGCACTATTT[C/T]TTCATGTGCAGGGCATCCTGGAACT
Phenotype
MIM: 606743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPALIN PubMed Links

Gene Details

Gene
OPALIN
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
There are no transcripts associated with this gene.

Gene
TLL2
Gene Name
tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012465.3 3283 Silent Mutation AAA,AAG K1014K NP_036597.1

View Full Product Details